Please login to your online account to display your discounted pricing

Promega™ Luciferase Assay System Components DFS Item


For use with Promega Luciferase Assay Systems (see cat. no. PRE1500)

Manufacturer: promega™  E1531

 View more versions of this product

Catalog No. PR-E1531

  • / Each
 

Description & Specifications

Specifications

Includes 1 x 30mL 5X Cell culture Lysis reagent
Quantity 30mL
Product Type Luciferase Cell Culture Lysis 5X Reagent
Concentration 5X
Storage Requirements -20°C
pH 7.8
Format Liquid