Please login to your online account to display your discounted pricing

Promega™ Luciferase Assay System Components


For use with Promega Luciferase Assay Systems (see cat. no. PRE1500)

Manufacturer: promega™  E3971

 View more versions of this product

Catalog No. PR-E3971

  • / Each
 

Description & Specifications

Specifications

Format Liquid
pH 7.6
Includes 1 x 30mL 5X Reporter Lysis Buffer
Concentration 5X
Quantity 30mL
Storage Requirements Room Temperature
Product Type Reporter Lysis 5X Buffer