missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ InsR Kinase Enzyme System DFS Item

Optimized for use with ADP-Glo™ Kinase Assay that measures ADP formed from kinase reaction

$588.50 - $588.50

Specifications

Storage Requirements Store the Kinase at -70°C and the rest of the components at -20°C
View More Specs

Includes: InsR Kinase Enzyme System (V3901)
  • Kinase: InsR, 10μg (Human, recombinant; amino acids 1011–end). MW: ∽70kDa
  • Substrate: Axltide (KKSRGDYMTMQIG); derived from the mouse Insulin receptor substrate 1 (amino acid 979–989)
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9411)
  • 1 x 10μg InsR Kinase Enzyme System (V901) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV3901 Promega™
V3901
10μg Each for $588.50
Specifications

Description

InsR is the insulin receptor tyrosine kinase that is involved in insulin signaling. Binding of insulin to the InsR stimulates glucose uptake. Insulin receptor signaling helps to maintain fuel homeostasis and prevent diabetes.

  • ADP-Glo Kinase assay is luminescent ADP detection assay
  • ADP is converted to ATP catalyzed by Ultra-Glo™ Luciferase that produces light
  • Luminescent signal correlates with ADP amount and kinase activity

Upon receipt, centrifuge the kinase and dispense it into smaller quantities.

Applications:

ADP-Glo™ Kinase Assay: For both primary screening as well as kinase selectivity profiling

Description & Specifications

Specifications

Store the Kinase at -70°C and the rest of the components at -20°C