Please login to your online account to display your discounted pricing

Promega™ InCELLect™ AKAP St-Ht31 Inhibitor Peptide


Cell-permeable peptides allow for in vivo studies of PKA localization

Manufacturer: promega™  V8221

 View more versions of this product

 Disclaimers

U.S. Pat. No. 6,610,657 and other patents pending.

Catalog No. PR-V8221

  • / Each
 

Description & Specifications

Specifications

Product Type AKAP St-Ht31 Control Peptide
Format Liquid
Molecular Weight 2797 ±3
Quantity 150μL
Storage Requirements -20°C
Purity >80%