Promega InCELLect™ AKAP St-Ht31 Inhibitor Peptide DFS Item
Cell-permeable peptides allow for in vivo studies of PKA localization
Manufacturer:  Promega V8211
 View more versions of this product
Catalog No. PR-V8211
Description
Description
  • Inhibits interaction between RII subunits of cAMP-Dependent Protein Kinase (PKA) and AKAP
  • Cell permeable, allowing for in vivo studies on PKA localization
  • Used with the InCELLect™ St-Ht31 Control Peptide to study the role of AKAP in mediating PKA responses
Specifications
Specifications
2797 ±3
150μL
10mM in 50mM Tris HCl (pH 7.5)
AKAP St-Ht31 Inhibitor Peptide
>80%
Liquid
−30 to −10°C
SDS
Documents
Promotions
Promotions
U.S. Pat. No. 6,610,657 and other patents pending.