Please login to your online account to display your discounted pricing

Promega™ InCELLect™ AKAP St-Ht31 Inhibitor Peptide

Cell-permeable peptides allow for in vivo studies of PKA localization

$541.42 - $541.42

Specifications

Quantity 150μL
Format Liquid
Molecular Weight 2797 ±3
Purity >80%
Storage Requirements -20°C
View More Specs

 Disclaimers

U.S. Pat. No. 6,610,657 and other patents pending.

Products
Catalog Number Mfr. No. Product Type Price Quantity    

PR-V8221

 
promega™
V8221
AKAP St-Ht31 Control Peptide Each for $541.42

PR-V8211

 
promega™
V8211
AKAP St-Ht31 Inhibitor Peptide Each for $541.42
Description & Specifications

Specifications

Quantity 150μL
Format Liquid
Molecular Weight 2797 ±3
Purity >80%
Storage Requirements -20°C

  • Stearated (St) forms of the peptide Ht-31
  • The presence of the hydrophobic stearated moiety enhances the cellular uptake of the peptide through the lipophilic microenvironment of the plasma membrane
  • Purity: >80% by HPLC

  • The inhibitor peptide has been shown to inhibit the interaction between the RII subunits of cAMP-Dependent Protein Kinase (PKA) and A-kinase anchoring protein (AKAP) in cell extracts
  • The control peptide, which has a very similar sequence but does not inhibit the interaction between protein kinase A and AKAP, has been used as a control peptide for studies with InCELLect AKAP St-Ht31 Inhibitor Peptide