Please login to your online account to display your discounted pricing

Promega™ ImProm-II™ Reverse Transcription System


Formulated for stand-alone applications or for easy transition to gene-specific target amplification

Manufacturer: promega™  A3800

 View more versions of this product

Catalog No. PR-A3800

  • / Each
 

Description & Specifications

Specifications

Includes Upstream control primer 100μL, Downstream control primer 100μL, MgCl2 1.2mL, Oligo(dT)15 Primer 50μg, dNTP mix 320μL, Random primers 50μg, 1.2kb kanamycin positive control RNA 5μg, ImProm-II 5X reaction buffer 600μL, ImProm-II reverse transcriptase 100μL, Recombinant RNasin ribonuclease inhibitor 2500U, Nuclease-free water 2 × 1250μL
Quantity 100 Reactions
Format Liquid
pH 7
For Use With (Application) Reverse transcription of RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
Product Type ImProm-II Reverse Transcription System