Please login to your online account to display your discounted pricing

Promega™ Tth DNA Polymerase


Available in a convenient, cost-effective concentration that is economical and easy to pipet

Manufacturer: promega™  M2105

 View more versions of this product

Catalog No. PR-M2105

  • / Each
 

Description & Specifications

Specifications

Inhibitors Recombinant RNasin™ Ribonuclease
Source Thermus thermophilus HB-8
pH 7.5
Includes 10X Reverse Transcription Buffer: 100mM Tris HCl (pH 8.3 at 25°C), 900mM KCl, 10X Chelate Buffer: 100mM Tris HCl (pH 8.3 at 25°C), 1M KCl, 7.5mM EGTA, 0.5% Tween™ 20, 50% Glycerol, Thermophilic DNA Pol
Quantity 500U
For Use With (Application) RT-PCR, PCR, primer extension, reverse transcription
Format Liquid
Concentration 5U/μL
Purity >95%
Molecular Weight 94000
Color Colorless
Storage Requirements -20°C
Format Liquid
Storage Buffer 10mM Tris HCl (pH 7.5 at 25°C), 300mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
Product Name Tth DNA Polymerase
Concentration 5U/μL
Quantity 500U