Please login to your online account to display your discounted pricing

Promega™ Tth DNA Polymerase


Available in a convenient, cost-effective concentration that is economical and easy to pipet

Manufacturer: promega™  M2101

 View more versions of this product

Catalog No. PR-M2101

  • / Each
 

Description & Specifications

Specifications

Inhibitors Recombinant RNasin™ Ribonuclease
pH 7.5
Molecular Weight 94000
Quantity 100U
Storage Buffer 10mM Tris HCl (pH 7.5 at 25°C), 300mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C
Format Liquid
Concentration 5U/μL
Color Colorless
Concentration 5U/μL
Purity >95%
Format Liquid
Product Name Tth DNA Polymerase
For Use With (Application) RT-PCR, PCR, primer extension, reverse transcription
Source Thermus thermophilus HB-8
Quantity 100U