Please login to your online account to display your discounted pricing

Promega™ Tth DNA Polymerase DFS Item

Available in a convenient, cost-effective concentration that is economical and easy to pipet

$102.98 - $424.00

Specifications

Color Colorless
Product Name Tth DNA Polymerase
For Use With (Application) RT-PCR, PCR, primer extension, reverse transcription
Inhibitors Recombinant RNasin™ Ribonuclease
Molecular Weight 94000
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PR-M2101

DFS Item
 
promega™
M2101
100U Each for $102.98

PR-M2105

DFS Item
 
promega™
M2105
500U Each for $424.00
Description & Specifications

Specifications

Color Colorless
Product Name Tth DNA Polymerase
For Use With (Application) RT-PCR, PCR, primer extension, reverse transcription
Inhibitors Recombinant RNasin™ Ribonuclease
Molecular Weight 94000
pH 7.5
Purity >95%
Source Thermus thermophilus HB-8
Storage Buffer 10mM Tris HCl (pH 7.5 at 25°C), 300mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements -20°C