Please login to your online account to display your discounted pricing

Promega™ Renilla Luciferase Assay System DFS Item


Fast and sensitive method of detecting the luciferase from sea pansy (Renilla reniformis)

Manufacturer: promega™  E2820

 View more versions of this product

Catalog No. PR-E2820

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
Quantity 1000 Assays
For Use With (Application) Detecting the Luciferase from Sea pansy (Renilla reniformis)