Please login to your online account to display your discounted pricing

Promega™ E. coli S30 T7 High-Yield Protein Expression System DFS Item


Express up to 500μg/mL of protein in 1 hour from plasmid vectors containing a T7 promoter and a ribosome binding site

Manufacturer: promega™  L1110

 View more versions of this product

 Disclaimers

U.S. Patents 5,292,658, and 5,418,155; Canadian Patent 2,105,984 and related patents.

Catalog No. PRL1110

  • / Each
 

Description & Specifications

Specifications

Product Type S30 T7 high yield Protein expression system
Yield 500μg/mL
Storage Requirements -70°C
Test Time 1 hr.
For Use With (Application) Protein-Protein interactions, Protein-DNA interactions, Enzyme activity, Antibody production
Quantity 24 Reactions