Promega™ Human Recombinant Tryptase DFS Item
Predominant protein in mast cell granules and cleaves proteins at arginine and lysine residues
Manufacturer:  Promega™ G5631
 View more versions of this product
Catalog No. PR-G5631
Specifications Description & Specifications
Specifications
100μg
2mg/mL
rhLung beta Tryptase
-20°C
High specific activity, consistent, safe, pure
1 x 100μg rhLung beta Tryptase