Please login to your online account to display your discounted pricing

Promega™ Human Recombinant Tryptase

Predominant protein in mast cell granules and cleaves proteins at arginine and lysine residues

$833.53 - $833.53

Specifications

Quantity 100μg
For Use With (Application) High specific activity, consistent, safe, pure
Storage Requirements -20°C
View More Specs
Products
Catalog Number Mfr. No. Concentration Formulation Product Type Price Quantity    

PR-G5631

 
promega™
G5631
2mg/mL 1 x 100μg rhLung beta Tryptase rhLung beta Tryptase Each for $833.53
Description & Specifications

Specifications

Quantity 100μg
For Use With (Application) High specific activity, consistent, safe, pure
Storage Requirements -20°C

  • High Specific Activity: Specific activity is consistently 130 to150% higher than native lung tryptase
  • Consistent: Recombinant protein expression results in uniform enzyme from batch to batch.
  • Safe: Void of human pathogens associated with native cadaveric tryptase
  • Pure: Skin β- and Lung β-Tryptase are free of other contaminating proteases, providing more active enzyme per milligram of protein and eliminating extraneous protein interactions observed with native tryptase