Please login to your online account to display your discounted pricing
Promega™ HisLink™ 96 Protein Purification System DFS Item
Provides a simple, quick and robust method of purifying multiple polyhistidine- or HQ-tagged expressed proteins from E. coli using a vacuum-based method
Manufacturer: promega™  V3680
 View more versions of this product
96-place vacuum manifold
 Disclaimers
Patent Pending.
This product is licensed for use under U.S. Pat. No. 6,174,704.
Certain applications of this product are covered by patents issued to parties other than Promega and applicable in certain countries. Because purchase of this product does not include a license to perform any of these patented applications, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.
Catalog No. PR-V3680
  • / Each
Description & Specifications
Specifications
Product Type HisLink™ 96 Protein Purification System
Storage Requirements 4°C
For Use With (Application) Quick and robust method of purifying multiple Polyhistidine-or HQ-tagged expressed Proteins from E. coli using a vacuum-based method
Quantity 1 x 96 Tests