Please login to your online account to display your discounted pricing

Promega™ HeLaScribe™ Nuclear Extract in vitro Transcription Grade


Derived from HeLa cell nuclei

Manufacturer: promega™  E3091

 View more versions of this product

Catalog No. PR-E3091

  • / Each
 

Description & Specifications

Specifications

Quantity 40 Reactions
For Use With (Application) Basal and transcription factor-regulated transcription assays, promoter strength assays and mapping of mRNA start sites
Product Type HeLaScribe Nuclear Extract, Transcription Grade
Storage Requirements Store at -70°C.