missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ HeLaScribe™ Nuclear Extract in vitro Transcription Grade DFS Item

Derived from HeLa cell nuclei

$407.67 - $1,067.86

Specifications

For Use With (Application) Basal and transcription factor-regulated transcription assays, promoter strength assays and mapping of mRNA start sites
Storage Requirements Store at -70°C.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-E3092 Promega™
E3092
160 Reactions Each for $1,067.86
PR-E3091 Promega™
E3091
40 Reactions Each for $407.67
Specifications

Description

  • Provide a cell-free system for in vitro transcription of eukaryotic genes

Description & Specifications

Specifications

Basal and transcription factor-regulated transcription assays, promoter strength assays and mapping of mRNA start sites
Store at -70°C.