Please login to your online account to display your discounted pricing

Promega™ HDAC-Glo™ I/II Assay and Screening Systems


A single-reagent-addition,homogeneous, luminescent assay that measures the relative activity of histone
deacetylase (HDAC) class I and II enzymes from cells, extracts or purified enzyme
sources.

Manufacturer: promega™  G6420

 View more versions of this product

Catalog No. PRG6420

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Determining HDAC inhibitor potency using purified Enzymes, extracts or cells directly in culture plates
pH 8
Storage Requirements -20°C
Kit Contents 1 x 25mL HDAC-Glo™ I or II Buffer, 1 x 1 vial HDAC-Glo™ I or II substrate, 1 x 10μL Developer Reagent, 1 x 10μL Trichostatin A
Type Assay
Quantity 10mL
Format Fluid