Please login to your online account to display your discounted pricing

Promega™ HB101 Competent Cells


Ready-to-use preparations provide convenient, efficient transformation

Manufacturer: promega™  L2011

 View more versions of this product

Catalog No. PR-L2011

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 200μL
Storage Requirements Store at -70°C.
pH 7.4
Includes HB101 Competent Cells (high efficiency) and Control DNA
Product Type Competent Cells
Format Fluid
Format Fluid
For Use With (Application) Transformation of unmethylated DNA derived from PCR, cDNA and many other sources
Concentration >108cfu/μg
Concentration >108cfu/μg
Product Name Competent Cells
Quantity 5 x 200μL