Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Vectors DFS Item


A variety vectors for mammalian, E. coli, and cell-free protein expression

Manufacturer: promega™  G2841

 View more versions of this product

Catalog No. PRG2841

  • / Each
 

Description & Specifications

Specifications

Quantity 20μg
Concentration 100ng/μL
Storage Requirements -20°C
Protein Expression Level Medium
For Use With (Application) Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression
Vector pFN22A HaloTag™ CMVd1 Flexi™ vector
Promoter CMVd1
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA