Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Vectors


A variety vectors for mammalian, E. coli, and cell-free protein expression

Manufacturer: promega™  G2681

 View more versions of this product

Catalog No. PRG2681

  • / Each
 

Description & Specifications

Specifications

Concentration 100ng/μL
Quantity 20μg
Vector pFN18K HaloTag™ T7 Flexi™ vector
Protein Expression Level High
Promoter T7
For Use With (Application) Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression
Storage Requirements -20°C
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA