Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Vectors


A variety vectors for mammalian, E. coli, and cell-free protein expression

Manufacturer: promega™  G2981

 View more versions of this product

Catalog No. PRG2981

  • / Each
 

Description & Specifications

Specifications

Vector pFN24K HaloTag™ CMVd3 Flexi™ vector
Promoter CMVd3
Storage Requirements -20°C
Quantity 20μg
For Use With (Application) Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression
Concentration 100ng/μL
Protein Expression Level Ultralow
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA