Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Vectors


A variety vectors for mammalian, E. coli, and cell-free protein expression

Manufacturer: promega™  G2881

 View more versions of this product

Catalog No. PRG2881

  • / Each
 

Description & Specifications

Specifications

Promoter CMVd3
Quantity 20μg
Storage Requirements -20°C
Protein Expression Level Ultralow
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA
Concentration 100ng/μL
Vector pFN24A HaloTag™ CMVd3 Flexi™ vector
For Use With (Application) Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression