Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Vectors


A variety vectors for mammalian, E. coli, and cell-free protein expression

Manufacturer: promega™  G1841

 View more versions of this product

Catalog No. PRG1841

  • / Each
 

Description & Specifications

Specifications

Vector pFN19K HaloTag™ T7 SP6 Flexi™ vector
Concentration 100ng/μL
Quantity 20μg
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA
For Use With (Application) Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression
Promoter Sp6 and T7
Storage Requirements -20°C