Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Vectors


A variety vectors for mammalian, E. coli, and cell-free protein expression

Manufacturer: promega™  G1611

 View more versions of this product

Catalog No. PRG1611

  • / Each
 

Description & Specifications

Specifications

Protein Expression Level Medium
Quantity 20μg
For Use With (Application) Cell imaging of Protein localization or trafficking in conjunction with the fluorescent HaloTag™ ligands by using expression of HaloTag™ fusion Proteins in E. coli and cell-free Protein expression
Vector pFC15A HaloTag™ CMVd1 Flexi™ vector
Concentration 100ng/μL
Storage Requirements -20°C
Promoter CMVd1
Storage Buffer 10mM Tris HCl (pH 8), 1mM EDTA