Learn More
Promega HaloTag™ Protein Purification System
HaloTag™ Protein Purification System is designed to purify proteins fused to the HaloTag™ novel protein tag that enhances expression and solubility of proteins from E. coli and enables the covalent, efficient and specific capture of a protein of interest.
Supplier: Promega G6270
Includes: Protein Purification System Sample Pack (Mfr. No. G6270) includes HaloLink Resin, 2.5mL of 25% slurry; HisLink Protein Purification Resin, 0.5mL; TEV Protease for use with HaloLink Resin, 200μL. Protein Purification System (G6280) includes HaloLink Resin, 25mL of 25% slurry; HisLink Protein Purification Resin, 5mL; TEV Protease for use with HaloLink Resin, 1.6mL.
Description
- Enhanced Expression and Solubility of Recombinant Proteins with HaloTag(R) Ligands
- Covalent, efficient and specific capture of your protein of interest
- Overcome limitations (i.e., poor capture of proteins expressed at low levels, protein loss during washing) associated with affinity tags
- Sample size is available (choose Cat.# G6270)
Specifications
COMPONENTS AT DIFFERENT TEMPS | |
HaloTag Protein Purification System | |
1/Each | |
HaloLink™ Resin, 2.5mL of 25% slurry, HisLink Protein purification Resin, 0.5mL, TEV Protease for use with HaloLink™ Resin, 200μL |
By use of this product, researcher agrees to be bound by the terms of this limited use statement. If the researcher is not willing to accept the conditions of this limited use statement, and the product is unused, Promega will accept return of the unused product and provide the researcher with a full refund.
Researchers may only use this product in their own research and may modify or link Promega HaloTag ligands to Promega or customer-supplied moieties. No other use or transfer of this product is authorized without the express, written consent of Promega including, without limitation, commercial use. With respect to any uses outside this label license, including any diagnostic, therapeutic or prophylactic uses, please contact Promega for supply and licensing information. Promega makes no representations or warranties of any kind, either expressed or implied, including for merchantability or fitness for a particular purpose with regards to the product. The terms of this agreement shall be governed under the laws of the State of Wisconsin, USA.
U.S. Patent 7,429,472 and other patents pending.
U.S. Patents 7,112,552; 7,304,015; 7,304,016; 7,354,750 and other patents pending.
Certain applications of this product may be covered by patents issued to parties other than Promega and applicable in certain countries. Because purchase of this product does not include a license to perform any of these patented applications, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.