missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Mammalian Pull-Down and Labeling System DFS Item

Allows the study of cellular localization, trafficking and protein turnover

$70.61 - $417.30

Specifications

Formulation Tris HCl 50mM, NaCl 150mM, Triton X-100 1% and Sodium Deoxycholate 0.1% (pH 7.5)
For Use With (Application) For study of cellular localization, trafficking and Protein turnover
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity pH Price Quantity  
Catalog Number Mfr. No. Quantity pH Price Quantity  
PRG9381 Promega™
G9381
40mL 7.5 Each for $70.61
PRG6591 Promega™
G6591
20μg 8 Each for $282.48
PRG6504 Promega™
G6504
24 Reactions 7.5 Each for $417.30
Specifications

Description

  • The HaloTag Mammalian Pull-Down Systems are designed to capture and purify intracellular binary and higher order protein complexes, including transient or weakly interacting partners.
  • Rapid, Efficient and Covalent Capture or Binary and Higher Order Complexes Directly from Lysates: Improved capture of protein partners, including transient interactions.
  • High Purity and Low Background: Improved accuracy in identification of proteins; covalent attachment allows bait protein to remain behind if desired.
  • Compatibility with All Downstream Methods or Analysis: Freedom to identify complexes in variety of applications including mass spectrometry.

Description & Specifications

Specifications

Tris HCl 50mM, NaCl 150mM, Triton X-100 1% and Sodium Deoxycholate 0.1% (pH 7.5)
For study of cellular localization, trafficking and Protein turnover