Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Mammalian Pull-Down and Labeling System


Allows the study of cellular localization, trafficking and protein turnover

Manufacturer: promega™  G6504

 View more versions of this product

Catalog No. PRG6504

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For study of cellular localization, trafficking and Protein turnover
Format Fluid
pH 7.5
Quantity 24 Reactions
Includes Buffers and Resin necessary to perform a HaloTag™ pull-down, G6504 Plus the HaloTag™ TMRDirect Ligand
Formulation Tris HCl 50mM, NaCl 150mM, Triton X-100 1% and Sodium Deoxycholate 0.1% (pH 7.5)