Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Interchangeable Labeling Technology


Multiple applications from one genetic construct: Add fluorescent labels, affinity handles or immobilize HaloTag fusion protein to a solid support

Manufacturer: promega™  G1002

 View more versions of this product

Catalog No. PRG1002

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Includes Fluorescent labels, affinity tags and attachments to a solid phase
Quantity 15μL
Format Liquid
Format Liquid
Product Type HaloTag Alexa Fluor 488 Ligand
Product Name HaloTag Alexa Fluor 488 Ligand
Quantity 15μL