Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Interchangeable Labeling Technology


Multiple applications from one genetic construct: Add fluorescent labels, affinity handles or immobilize HaloTag fusion protein to a solid support

Manufacturer: promega™  G1001

 View more versions of this product

Catalog No. PRG1001

  • / Each
 

Description & Specifications

Specifications

Quantity 30μL
For Use With (Application) Protein localization, Protein trafficking, Protein capture, cell to Gel analysis, Protein to Protein interactions
Product Type HaloTag Alexa Fluor 488 Ligand
Includes Fluorescent labels, affinity tags and attachments to a solid phase
Format Fluid