Promega™ HaloTag™ Interchangeable Labeling Technology DFS Item
The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.
Manufacturer:  Promega™ G8582
 View more versions of this product
Catalog No. PRG8582
Description
Description
The HaloTag Technology is a platform technology for enabling covalent protein labeling and immobilization in vivo and in vitro. Products based on the HaloTag Technology enable researchers to study protein function in a biochemical and cellular environment. The technology is based on the efficient formation of a covalent bond between a specially designed reporter protein encoded by a HaloTag Vector and a specific ligand in living cells, in solution or on a solid support.
  • Create a HaloTag fusion protein with one easy cloning step
  • HaloTag protein creates strong and very specific covalent bonds with the different HaloTag ligands
  • Cell membrane permeable fluorescent ligands allow imaging of live or fixed cells
  • HaloTag PEG-Biotin Ligand for pull-down applications
Protein localization, Protein trafficking, Protein capture, Cell-to-gel analysis, Protein:protein interactions
Specifications
Specifications
Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Fluorescent labels, affinity tags and attachments to a solid phase
15μL
Fluid
HaloTag Coumarin Ligand
−30 to −10°C