Please login to your online account to display your discounted pricing

Promega™ HaloTag™ Interchangeable Labeling Technology


Multiple applications from one genetic construct: Add fluorescent labels, affinity handles or immobilize HaloTag fusion protein to a solid support

Manufacturer: promega™  G2991

 View more versions of this product

Catalog No. PRG2991

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Format Fluid
Format Fluid
Includes Fluorescent labels, affinity tags and attachments to a solid phase
Product Type HaloTag TMRDirect Ligand
Quantity 30μL
Product Name HaloTag TMRDirect Ligand
Quantity 30μL