Promega HaloTag™ Interchangeable Labeling Technology DFS Item
The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.
Supplier:  Promega G8272
 View more versions of this product
Catalog No. PRG8272
Description
Description
  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling
Specifications
Specifications
-30°C to -10°C
Fluorescent labels, affinity tags and attachments to a solid phase
30 μL
Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
HaloTag diAcFAM Ligand
Fluid