Promega HaloTag™ Interchangeable Labeling Technology
The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.
Supplier: Promega G8252
Description
- Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
- Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
- Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling
Specifications
-30°C to -10°C | |
Fluorescent labels, affinity tags and attachments to a solid phase | |
15 μL |
Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions | |
HaloTag TMR Ligand | |
Fluid |