Promega HaloTag™ Interchangeable Labeling Technology DFS Item
The HaloTag™ Technology enables covalent protein labeling and immobilization in vivo and in vitro.
Manufacturer:  Promega G2802
 View more versions of this product
Catalog No. PRG2802
Description
Description
  • Covalently Attach Fluorescent Labels or Affinity Tags to Membrane or Intracellular Proteins
  • Cell permeant TMR, Oregon Green(R), diAcFAM and Coumarin ligands readily cross the cell membrane for labeling intracellular proteins
  • Cell impermeant tags include Alexa Fluor(R) 488 and 660 for quick cell surface labeling
Specifications
Specifications
Protein localization, Protein trafficking, Protein capture, Cell to Gel analysis, Protein to Protein interactions
Fluorescent labels, affinity tags and attachments to a solid phase
15μL
Fluid
HaloTag Oregon Green Ligand
−30 to −10°C