missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega HaloTag™ Fusion (N-Terminal) Mammalian Expression Vectors DFS Item

Vectors designed for expression of N-terminal-tagged HaloTag™ fusion proteins in mammalian cells. Once expressed, use the HaloTag™ fusion protein for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag™ Ligands.

$319.28 - $319.28

Specifications

Promoter CMV
For Use With (Application) Transient and stable gene expression, cell imaging of Protein localization or trafficking in conjunction
Protein Expression Level High
Quantity 20μg
Storage Requirements −30 to −10°C
View More Specs

Products 1
Catalog Number Mfr. No. Concentration Storage Buffer Vector Price Quantity  
Catalog Number Mfr. No. Concentration Storage Buffer Vector Price Quantity  
PRG7721 View Documents Promega™
G7721*
1μg/μL 10mM Tris HCl (pH 7.4), 1mM EDTA pHTN HaloTag™ CMV-neo Flexi™ vector Each for $319.28
 
Description

Description

  • Append N-Terminal Tag for High-Level Protein Expression in Mammalian Cells
  • Enables constitutive protein expression in mammalian cells
  • Includes CMV immediate-early enhancer/promoter
  • Suited to stable or transient gene expression
Specifications

Specifications

CMV
High
−30 to −10°C
Transient and stable gene expression, cell imaging of Protein localization or trafficking in conjunction
20μg