Please login to your online account to display your discounted pricing

Promega™ HaloLink™ Resins


Solid support that allows covalent and oriented attachment of HaloTag™ fusion proteins to a Sepharose™ surface

Manufacturer: promega™  G1912

 View more versions of this product

Catalog No. PRG1912

  • / Each
 

Description & Specifications

Specifications

Sufficient For Covalent and oriented attachment of HaloTag™ fusion Proteins to a solid surface
Product Type HaloLink™ Resin
Storage Requirements 4°C
Quantity 1.25mL