missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ HaloLink™ Resins DFS Item

Solid support that allows covalent and oriented attachment of HaloTag™ fusion proteins to a Sepharose™ surface

$141.24 - $470.80

Specifications

Product Type HaloLink™ Resin
Storage Requirements 4°C
Sufficient For Covalent and oriented attachment of HaloTag™ fusion Proteins to a solid surface
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRG1912 Promega™
G1912
1.25mL Each for $141.24
PRG1914 Promega™
G1914
10mL Each for $470.80
PRG1913 Promega™
G1913
2.5mL Each for $204.38
Specifications

Description

  • Covalent Attachment: Enables stringent washing, minimizing nonspecific background without dissociation of bound HaloTag Fusion proteins
  • -Fast Binding Kinetics: Enhances the detection of protein: protein interaction and enables binging of proteins at low concentrations.
  • -Oriented Immobilization: Allows maximal enzyme activity of bound protein.
  • -High Binding Capacity: One millimeter of settled resin binds >7mg of HaloTag fusion proteins.

Description & Specifications

Specifications

HaloLink™ Resin
4°C
Covalent and oriented attachment of HaloTag™ fusion Proteins to a solid surface