Please login to your online account to display your discounted pricing

Promega™ Growth Factors DFS Item


Obtained from recombinant human DNA expressed in E. coli (rh), male mouse submaxillary glands (m), or natural human sources

Manufacturer: promega™  G5241

 View more versions of this product

Catalog No. PR-G5241

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity E. coli
Source Human
For Use With (Application) Biological effects of this molecule include induction of apoptosis, cytolysis or cytostasis of tumor cells
Type rhTNF-alpha
Molecular Weight 17
Quantity 10μg