Please login to your online account to display your discounted pricing

Promega™ Growth Factors


Obtained from recombinant human DNA expressed in E. coli (rh), male mouse submaxillary glands (m), or natural human sources

Manufacturer: promega™  G5021

 View more versions of this product

Catalog No. PR-G5021

  • / Each
 

Description & Specifications

Specifications

Type Nerve Growth Factor
Cross Reactivity E. coli
Lyophilized Yes
Stability 6 Months
Molecular Weight 6.2
Source Human
Quantity 100μg
Storage Requirements -20°C