missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ Growth Factors DFS Item

Obtained from recombinant human DNA expressed in E. coli (rh), male mouse submaxillary glands (m), or natural human sources

$248.00 - $464.00

Specifications

Cross Reactivity E. coli
Source Human
View More Specs

Products 3
Catalog Number Mfr. No. Type Molecular Weight Quantity Price Quantity  
Catalog Number Mfr. No. Type Molecular Weight Quantity Price Quantity  
PR-G5021 View Documents Promega™
G5021
Nerve Growth Factor 6.2 100μg Each for $248.00
PR-G5071 View Documents Promega™
G5071
Epidermal Growth Factor 17.5 25μg Each for $457.00
PR-G5241 View Documents Promega™
G5241
rhTNF-alpha 17 10μg Each for $464.00
Description

Description

  • Activity is expressed as the effective dose (ED50) or inhibitory concentration (IC50)

1At least half the activity (ED50) compared to International Reference Standard, where available.

Specifications

Specifications

E. coli
Human