Please login to your online account to display your discounted pricing

Promega™ GoTaq™ qPCR Master Mix DFS Item


Provides robust real-time qPCR with high levels of reliability, reproducibility and sensitivity

Manufacturer: promega™  A6002

 View more versions of this product

Catalog No. PRA6002

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Dye-Based Detection, qPCR and RT-qPCR
Storage Requirements -20°C
Volume 1000 x 50μL reactions
No. of Reactions 1000.0
Includes 5 x (5 x 1mL GoTaq™ qPCR master mix, 2X, 1 x 100μL CXR reference dye, 2 x 13mL Nuclease free Water)
Concentration 2X