Please login to your online account to display your discounted pricing

Promega™ GoTaq™ qPCR Master Mix DFS Item


Provides robust real-time qPCR with high levels of reliability, reproducibility and sensitivity

Manufacturer: promega™  A6001

 View more versions of this product

Catalog No. PRA6001

  • / Each
 

Description & Specifications

Specifications

Concentration 2X
Storage Requirements -20°C
For Use With (Application) Dye-Based Detection, qPCR and RT-qPCR
Includes 5 x 1mL GoTaq™ qPCR master mix, 2X, 1 x 100μL CXR reference dye, 2 x 13mL Nuclease free Water
No. of Reactions 200.0
Volume 200 x 50μL reactions