Please login to your online account to display your discounted pricing

Promega™ GoTaq™ PCR Core Systems


Provide a set of performance-tested PCR reagents

Manufacturer: promega™  M7665

 View more versions of this product

Catalog No. PR-M7665

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) PCR, RT-PCR
Includes 1 x 100μL Upstream control primer, 1 x 100μL downstream control primer, 1 x 1.2mL MgCl2, 1 x 200μL PCR nucleotide mix, 1 x 100μL positive control plasmid DNA, 1 x 250u GoTaq™
No. of Reactions 200.0
Storage Requirements -30 to -10°C