missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega GoTaq™ PCR Core Systems DFS Item

Provide a set of performance-tested PCR reagents

$151.00 - $166.00

Specifications

For Use With (Application) PCR, RT-PCR
No. of Reactions 200.0
Storage Requirements -30 to -10°C
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
PR-M7660 View Documents Promega™
M7660
1 x 1.2mL MgCl2, 1 x 200μL PCR nucleotide mix, 1 x 250μL GoTaq™ DNA Polymerase, 2 x 1mL 5X colorless GoTaq™ flexi Buffer, 2 x 1mL 5X green GoTaq™ flexi Buffer Each for $151.00
PR-M7665 View Documents Promega™
M7665
1 x 100μL Upstream control primer, 1 x 100μL downstream control primer, 1 x 1.2mL MgCl2, 1 x 200μL PCR nucleotide mix, 1 x 100μL positive control plasmid DNA, 1 x 250u GoTaq™ Each for $166.00
Description

Description

  • With optimization tools for reaction flexibility
  • PCR Core System I includes basic PCR reaction components
  • PCR Core System II also includes positive controls to help troubleshoot PCR parameters

Specifications

Specifications

PCR, RT-PCR
200.0
-30 to -10°C