Promega™ GoTaq™ Hot Start Polymerase: Colorless Master Mix, 2X DFS Item
GoTaq™ Hot Start Master Mixes are all-in-one master mixes (just add template and primers) that contain the high-performance GoTaq™ DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two
Manufacturer:  Promega™ M5132
 View more versions of this product
Catalog No. PR-M5132
Description
Description
  • Proprietary antibody bound to the polymerase blocks polymerase activity and permits room-temperture setup, but only requires a two-minute activation at 94°-95°C
  • GoTaq Flexi Reaction Buffers contain a compound that increases sample density and serves as both reaction buffer and gel-loading solution, saving time in post-PCR anaylsis
  • Colorless GoTaq Flexi Buffer is used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction
Hot-start PCR, complex genomic templates, T-vector cloning, two-step RT-PCR
Specifications
Specifications
2 min.
2X Hot start colorless master mix (2 x 1250μL), Nuclease free Water (2 x 1250μL)
-20°C
Hot-start PCR, complex genomic templates, T-vector cloning, two-step RT-PCR
100.0
5U/μL