Please login to your online account to display your discounted pricing

Promega™ GoTaq™ Hot Start Polymerase: Colorless Master Mix, 2X

Combines the robust amplification and sensitivity of GoTaq DNA Polymerase with hot-start technology to deliver even greater specificity

$105.00 - $839.00

Specifications

Activation Time 2 min.
Concentration 5U/μL
For Use With (Application) Hot-start PCR, complex genomic templates, T-vector cloning, two-step RT-PCR
Includes 2X Hot start green master mix (1 x 25mL), Nuclease free Water (1 x 25mL)
Storage Requirements -20°C
View More Specs
Includes:  GoTaq Hot Start Polymerase, buffer, dNTPs, and MgCl2, and supplied with nuclease-free water.
Products
Catalog Number Mfr. No. No. of Reactions Price Quantity    

PR-M5132

 
promega™
M5132
100.0 Each for $105.00

PR-M5133

 
promega™
M5133
1000.0 Each for $839.00
Description & Specifications

Specifications

Activation Time 2 min.
Concentration 5U/μL
For Use With (Application) Hot-start PCR, complex genomic templates, T-vector cloning, two-step RT-PCR
Includes 2X Hot start green master mix (1 x 25mL), Nuclease free Water (1 x 25mL)
Storage Requirements -20°C

  • Proprietary antibody bound to the polymerase blocks polymerase activity and permits room-temperture setup, but only requires a two-minute activation at 94°-95°C
  • GoTaq Flexi Reaction Buffers contain a compound that increases sample density and serves as both reaction buffer and gel-loading solution, saving time in post-PCR anaylsis
  • Colorless GoTaq Flexi Buffer is used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction

Hot-start PCR, complex genomic templates, T-vector cloning, two-step RT-PCR

U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.