missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega GoTaq™ Hot Start Polymerase: Colorless Master Mix, 2X
GoTaq™ Hot Start Master Mixes are all-in-one master mixes (just add template and primers) that contain the high-performance GoTaq™ DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two
Supplier: Promega M5133
Includes: GoTaq Hot Start Polymerase, buffer, dNTPs, and MgCl2, and supplied with nuclease-free water.
Description
- Hot-Start GoTaq(R) DNA Polymerase in a Ready-to-Use Master Mix
- All-in-one master mix: just add template and primers
- Minimize nonspecific amplification and primer-dimers
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.
Specifications
2X Hot start green master mix (1 x 25mL), Nuclease free Water (1 x 25mL). Store at -20°C | |
2 min. | |
1000 |
Cloning | |
5U/μL |