Please login to your online account to display your discounted pricing

Promega™ GoScript™ Reverse Transcription System


Combines a reverse transcriptase with a specialized set of reagents for efficient synthesis of first-strand cDNA optimized for QPCR amplification

Manufacturer: promega™  A5000

 View more versions of this product

 Disclaimers

U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.
U.S. Patent 5,552,302, Australian Patent 646803 and other patents.

Catalog No. PRA5000

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) RT-PCR, RT-QPCR
Includes 1 x 750μL MgCl2, 1 x 300μL GoScript™ 5X reaction Buffer, 1 x 50μL GoScript™ reverse Transcriptase, 1 x 50μg oligo(dT)15 primer, 1 x 200
No. of Reactions 50.0
Storage Requirements -20°C