Please login to your online account to display your discounted pricing

Promega™ Go Taq™ Master Mixes


Save time during reaction setup and PCR product analysis

Manufacturer: promega™  M7122

 View more versions of this product

Catalog No. PR-M7122

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) PCR product analysis
No. of Reactions 100.0
Concentration 2X
Storage Requirements -20°C
Includes 2 x 1250μL GoTaq™ Green MasterMix, 2X, 2 x 1250μL Nuclease free Water, blue dye and a yellow dye, GoTaq DNA Polymerase, dNTPs, MgCl2 and reaction Buffers in a single tube